missing translation for 'onlineSavingsMsg'
Learn More

Calcium (-)-Hydroxycitrate, TRC

Product Code. 30342228
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342228 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342228 Supplier Toronto Research Chemicals Supplier No. TRCC14560010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Hydroxycitric Acid Calcium Salt
CAS 921226-01-9
Molecular Formula 2 C6 H5 O8 . 3 Ca
Quantity 10 mg
Synonym D-erythro-Pentaric acid, 3-C-carboxy-2-deoxy-, calcium salt (2:3), Calcium (-)-hydroxycitrate
InChI Formula InChI=1S/2C6H8O8.3Ca/c2*7-2(8)1-6(14,5(12)13)3(9)4(10)11;;;/h2*3,9,14H,1H2,(H,7,8)(H,10,11)(H,12,13);;;/q;;3*+2/p-6/t2*3-,6+;;;/m11.../s1
SMILES [Ca+2].[Ca+2].[Ca+2].O[C@H](C(=O)[O-])[C@@](O)(CC(=O)[O-])C(=O)[O-].O[C@H](C(=O)[O-])[C@@](O)(CC(=O)[O-])C(=O)[O-]
IUPAC Name tricalcium;(1S,2S)-1,2-dihydroxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol) 530.43
Formula Weight 529.8847
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.