missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Hydrate, TRC

Product Code. 30138293
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138293 100 g 100g
30133885 50 g 50g
30012125 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138293 Supplier Toronto Research Chemicals Supplier No. TRCC145060100G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula Ca . 2 Cl . 2 H2 O
Molecular Weight (g/mol) 147.01
Synonym Calcium Chloride Dihydrate
IUPAC Name calcium;dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Ca+2]

Specifications

Quantity 100 g
InChI Formula InChI=1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2
Formula Weight 145.9214
Chemical Name or Material Calcium chloride dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.