missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride (Anhydrous), TRC

Product Code. 30137993
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137993 50 g 50g
30135994 100 g 100g
30007947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137993 Supplier Toronto Research Chemicals Supplier No. TRCC14505550G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula Ca . 2 Cl
Molecular Weight (g/mol) 110.98
Synonym Calcium chloride (CaCl2), Calcium chloride (8CI), Bovikalc, CalPlus, Calcium dichloride, Calcium(2+) chloride, Calcosan, Calmate R, Calol, Calzina oral, Chrysoxel C 4, Daraccel, Dowflake, Intergravin-orales, Kalgan, Liquidow, Peladow, Stopit, U-Ramin MC, Calcium Chloride anhydrous
IUPAC Name calcium;dichloride
SMILES [Cl-].[Cl-].[Ca+2]

Specifications

Quantity 50 g
InChI Formula InChI=1S/Ca.2ClH/h;2*1H/q+2;;/p-2
Formula Weight 109.9003
Chemical Name or Material Calcium Chloride anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.