missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, TRC

Product Code. 30343353
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Unit Size:
250g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343353 5 g 5g
30334724 250 g 250g
30331729 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30343353 Supplier Toronto Research Chemicals Supplier No. TRCC1450505G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Carbonate (AS)
CAS 471-34-1
Molecular Formula CCaO3
Quantity 5 g
SMILES [Ca+2].[O-]C(=O)[O-]
Molecular Weight (g/mol) 100.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.