Learn More
Calcifediol-d6, MedChemExpress
Description
Calcifediol-d6 is the deuterium labeled Calcifediol. Calcifediol, a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.
Specifications
Specifications
| Chemical Name or Material | Calcifediol-d6 |
| CAS | 78782-98-6 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C27H38D6O2 |
| Color | White |
| SMILES | O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2 |
| Industry Type | Metabolism-sugar/lipid metabolism |
| Molecular Weight (g/mol) | 406.67 |
| Quantity | 1 mg |
| Show More |