missing translation for 'onlineSavingsMsg'
Learn More

Caffeidine, TRC

Product Code. 30010423
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010423 100 mg 100mg
30007670 500 mg 500mg
30007154 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010423 Supplier Toronto Research Chemicals Supplier No. TRCC080200100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20041-90-1
Molecular Formula C7 H12 N4 O
Molecular Weight (g/mol) 168.2
Synonym N,1-Dimethyl-4-(methylamino)imidazole-5-carboxamide, Imidazole-5-carboxamide, N,1-dimethyl-4-(methylamino)- (8CI), 1H-Imidazole-5-carboxamide, N,1-dimethyl-4-(methylamino)-
IUPAC Name N,3-dimethyl-5-(methylamino)imidazole-4-carboxamide
SMILES CNC(=O)c1c(NC)ncn1C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H12N4O/c1-8-6-5(7(12)9-2)11(3)4-10-6/h4,8H,1-3H3,(H,9,12)
Formula Weight 168.1011
Percent Purity >95%
Chemical Name or Material Caffeidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.