missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ E-Cadherin Rabbit Monoclonal Antibody

Ensure accurate and reliable results in immunohistochemistry applications with Epredia™ Cadherin E, Rabbit Monoclonal Antibody.

165.00 EUR - 798.25 EUR

Specifications

Antigen E-Cadherin
Clone EP700Y
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12653597
View Documents
Epredia™
RM-2100-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12643597
View Documents
Epredia™
RM-2100-S
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
892.00 EUR
798.25 EUR / 1mL
Save 93.75 EUR
10% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12633597
View Documents
Epredia™
RM-2100-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
557.00 EUR
507.71 EUR / 7mL
Save 49.29 EUR
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12623597
View Documents
Epredia™
RM-2100-RQ
12 mL Ready-to-Use
ONLINE EXCLUSIVE PRICE
727.00 EUR
649.33 EUR / 12mL
Save 77.67 EUR
10% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominantly in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis. E-cadherin is used in distinguishing LCIS from DCIS and that of ILC from IDC in indeterminate cases.

Host Species: Rabbit

Clone: EP700Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from the fifth cadherin domain of human cadherin

Molecular Weight: 135/80kDa

Positive Control: Breast Carcinoma

Cellular Localization: Cell Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

E-Cadherin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
A synthetic peptide derived from the fifth cadherin domain of human cadherin
EP700Y
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.