missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ E-Cadherin Rabbit Monoclonal Antibody

Ensure accurate and reliable results in immunohistochemistry applications with Epredia™ Cadherin E, Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-2100-S0

 View more versions of this product

Product Code. 12653597

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominantly in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis. E-cadherin is used in distinguishing LCIS from DCIS and that of ILC from IDC in indeterminate cases.

Host Species: Rabbit

Clone: EP700Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from the fifth cadherin domain of human cadherin

Molecular Weight: 135/80kDa

Positive Control: Breast Carcinoma

Cellular Localization: Cell Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

E-Cadherin
Monoclonal
Unconjugated
A synthetic peptide derived from the fifth cadherin domain of human cadherin
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
EP700Y
Rabbit
100 μL
Cancer and Tumor Biology
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.