Learn More
Cadazolid, MedChemExpress
Product Code. 16443018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443018 10 mg 10mg
16434326 100 mg 100mg
16429586 50 mg 50mg
16425757 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16443018 Supplier Medchem Express Supplier No. HY100436
 Restricted Item
Estimated Shipment: 02-11-2026
MedChemExpress Cadazolid (ACT-179811) is a new oxazolidinone antibiotic with potent activity against Clostridium difficile.
Cadazolid (ACT-179811) is a new oxazolidinone antibiotic with potent activity against Clostridium difficile.
Specifications
Chemical Name or Material Cadazolid
Physical Form Solid
CAS 1025097-10-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C29H29F2N3O8
For Use With (Application) COVID-19-immunoregulation
Synonym ACT-179811
Solubility Information DMSO : 100 mg/mL (170.78 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4CCC(O)(COC5=CC=C(N6C(O[C@@H](CO)C6)=O)C=C5F)CC4)=C3)C1=O)O
Molecular Weight (g/mol) 585.55
Formula Weight 585.55
Percent Purity 98.66%
Grade Research