Learn More
Cadaverine, TRC
Product Code. 30137424
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137424 100 g 100g
30136986 25 g 25g
30006174 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137424 Supplier Toronto Research Chemicals Supplier No. TRCC058000100G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 462-94-2
Molecular Formula C5 H14 N2
Molecular Weight (g/mol) 102.18
Synonym 1,5-Amylene diamine, 1,5-Diamino-n-pentane, 1,5-Diaminopentane, 1,5-Pentamethylenediamine, Cadaverine, Pentamethylenediamine, α,ω-Pentanediamine
IUPAC Name pentane-1,5-diamine
SMILES NCCCCCN
Specifications
Quantity 100 g
InChI Formula InChI=1S/C5H14N2/c6-4-2-1-3-5-7/h1-7H2
Formula Weight 102.1157
Chemical Name or Material 1,5-Diaminopentane