missing translation for 'onlineSavingsMsg'
Learn More

Cabotegravir-d3, TRC

Product Code. 30335982
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335982 5 mg 5mg
30335852 1 mg 1mg
30332628 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335982 Supplier Toronto Research Chemicals Supplier No. TRCC0501025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cabotegravir-d3
Purity Grade Notes HPLC
Molecular Formula C19H14D3F2N3O5
Quantity 5 mg
Synonym (3S,11aR)-N-[(2,4-difluorophenyl)methyl]-2,3,5,7,11,11a-hexahydro-6-hydroxy-3-methyl-5,7-dioxo-Oxazolo[3,2-a]pyrido[1,2-d]pyrazine-8-carboxamide-d3
InChI Formula InChI=1S/C19H17F2N3O5/c1-9-8-29-14-7-23-6-12(16(25)17(26)15(23)19(28)24(9)14)18(27)22-5-10-2-3-11(20)4-13(10)21/h2-4,6,9,14,26H,5,7-8H2,1H3,(H,22,27)/t9-,14+/m0/s1/i1D3
SMILES O=C(C1=CN(C2=C(O)C1=O)C[C@](OC[C@@H]3C([2H])([2H])[2H])([H])N3C2=O)NCC4=CC=C(F)C=C4F
IUPAC Name (3R,6S)-N-[(2,4-difluorophenyl)methyl]-10-hydroxy-8,11-dioxo-6-(trideuteriomethyl)-4-oxa-1,7-diazatricyclo[7.4.0.03,7]trideca-9,12-diene-12-carboxamide
Molecular Weight (g/mol) 408.37
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.