missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CABLES1 Polyclonal antibody specifically detects CABLES1 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CABLES1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000 |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | CABL1, CABLES, Cdk5 and Abl enzyme substrate 1, CDK5 and ABL1 enzyme substrate 1, FLJ35924, HsT2563, IK3-1, Interactor with CDK3 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-368 of human CABLES1 (NP_612384.1). MLSKRGCHARIYADFPIRRLISQRSSLETLEDIEENAPLRRCRTLSGSPRPKNFKKIHFIKNMRQHDTRNGRIVLISGRRSFCSIFSVLPYRDSTQVGDLKLDGGRQSTGAVSLKEIIGLEGVELGADGKTVSYTQFLLPTNAFGARRNTIDSTSSFSQFRNLSHRSLSIGRASGTQGSLDTGSDLGDFMDYDPNLLDDPQWPCGKHKRVLIFPSYMTTVIDYVKPSDLKKDMNETFKEKFPHIKLTLSKIRSLKREMRKLAQEDCGLEEPTVAMAFVYFEKLALKGKLNKQNRKLCAGACVLLAAKIGSDLKKHEVKHLIDKLEEKFRLNRRELIAFEFPVLVALEFALHLPEHEVMPHYRRLVQSS |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?