Learn More
Hirschmann™ Cable computer connection RS232
Product Code. 10367301
Change view
Click to view available options
For Use With (Equipment):
For opus™
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10367301 For opus™ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10367301 Supplier Hirschmann™ Supplier No. 9564005
  • ONLINE EXCLUSIVE PRICE
    168.00 EUR
    107.00 EUR / Pack of 1
    Save 61.00 EUR
    36% Off
Estimated Shipment: 16-12-2026
Computer connection cable with RS232 connector.
  • For computer controlled operation
  • RS232
  • For opus™
Specifications
Description Serial interface connecting cable
For Use With (Equipment) For opus™
Connector Types RS232
Length (Metric) 3 m