Learn More
Medline Scientific™ C-Mount Fittings
Product Code. 12673025
Change view
Click to view available options
For Use With (Equipment):
Magnum and Reflex Series microscopes
Varizoom series microscopes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12673025 Varizoom series microscopes Each
12413358 Magnum and Reflex Series microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12673025 Supplier Medline Scientific™ Supplier No. 7700.2345
  • ONLINE EXCLUSIVE PRICE
    183.00 EUR
    173.84 EUR / Pack of 1
    Save 9.16 EUR
    5% Off
Estimated Shipment: 16-02-2027
For connecting the camera to the microscope, ensuring that parfocality is retained by placing the chamera chip in the same focal plan as the eyepieces so that no refocusing is required when switching between manual or on-screen viewing.
  • Each C mount is specific to the relevant microscope series
  • Available for Varizoom series, Magnum and Reflex series, and Triton II and Inverso TC series
Specifications
Type C-mount Adapter
For Use With (Equipment) Varizoom series microscopes
Quantity 1 Ea.