Learn More
Masterflex™ C-Flex™ Tubing
Use this tubing for general laboratory, biotech and pharmaceutical applications requiring high performance. Cole-Parmer™ C-Flex Tubing features exceptional tensile and tear strength, good biocompatibility an excellent chemical resistance.
Brand:  Masterflex™ 06422-11
Product Code. 12428640
  • 127.00 EUR / 1 rolls
Only null left
Estimated Shipment: 03-08-2026
Description
Transparent tubing has low gas permeability and a smooth surface. Provides an economical alternative to platinum silicone. Product is manufactured in accordance with quality regulations CFR Title 21, Part 820 Quality System Requirements (GMP).
  • Temperature range: ndash;40 to 275°C
  • Vacuum service of 760mm Hg at 23°C
  • Sterilize: Autoclave or ethylene oxide