missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone, Sigma-Aldrich™

Product Code. 15892331
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15892331 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15892331 Supplier Sigma Aldrich Supplier No. 8417780025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 495-40-9
Melting Point 11°C to 13°C
Density 0.988 g/cm3
Boiling Point 220°C to 222°C/1013 hPa
MDL Number MFCD00009397
Quantity 25 mL
Synonym Butyrophenone, Butyrophenone
Molecular Weight (g/mol) 148.2
Grade Research
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.