Learn More
Butyraldoxime, TRC
Specifications
Specifications
| Chemical Name or Material | Butyraldoxime |
| CAS | 110-69-0 |
| Molecular Formula | C4H9NO |
| Quantity | 5 g |
| Synonym | Butanaldoxime, Butylaldoxime, Butyraldoxime, NSC 1487, n-Butylaldoxime, n-Butyraldoxime |
| InChI Formula | InChI=1S/C4H9NO/c1-2-3-4-5-6/h4,6H,2-3H2,1H3 |
| SMILES | CCCC=NO |
| IUPAC Name | N-butylidenehydroxylamine |
| Molecular Weight (g/mol) | 87.12 |
| Formula Weight | 87.0684 |