Learn More
Butyl 2-[[3-(Trifluoromethyl)phenyl]-amino]benzoate (Butyl Flufenamate), Mikromol™
Beschreibung
Butyl 2-[[3-(Trifluoromethyl)phenyl]-amino]benzoate (Butyl Flufenamate)
Spezifikation
Spezifikation
| Chemical Name or Material | Butyl 2-[[3-(Trifluoromethyl)phenyl]-amino]benzoate (Butyl Flufenamate) |
| CAS | 67330-25-0 |
| Analyte or Component Names | Butyl 2-[[3-(Trifluoromethyl)phenyl]-amino]benzoate (Butyl Flufenamate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 F3 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Butyl 2-[[3-(Trifluoromethyl)phenyl]-amino]benzoate, Etofenamate Imp. B (EP), Butyl Flufenamate, Etofenamate Impurity B |
| InChI Formula | InChI=1S/C18H18F3NO2/c1-2-3-11-24-17(23)15-9-4-5-10-16(15)22-14-8-6-7-13(12-14)18(19,20)21/h4-10,12,22H,2-3,11H2,1H3 |
| SMILES | CCCCOC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F |
| IUPAC Name | butyl 2-[3-(trifluoromethyl)anilino]benzoate |
| Mehr anzeigen |