missing translation for 'onlineSavingsMsg'
Learn More

Butane-1-sulfonic acid, LiChropur™ EMD-Millipore

Product Code. 15815890
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15815890 25 g Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815890 Supplier Supelco Supplier No. 1183030025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ion pair reagents for analytical HPLC LiChropur

TRUSTED_SUSTAINABILITY

Specifications

CAS 2386-54-1
Packaging Glass
Molecular Formula C4H9NaO3S
MDL Number MFCD00007540
Quantity 25 g
Synonym sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
SMILES [Na+].CCCCS([O-])(=O)=O
IUPAC Name sodium butane-1-sulfonate
Molecular Weight (g/mol) 160.16
PubChem CID 4096517
Formula Weight 160.17
Grade Not Available
Chemical Name or Material Butane-1-sulfonic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.