missing translation for 'onlineSavingsMsg'
Learn More

Buspirone Hydrochloride, Mikromol™

Product Code. 18090880
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090880 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18090880 Supplier Mikromol Supplier No. MM0464.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Buspirone Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buspirone hydrochloride
CAS 33386-08-2
Analyte or Component Names Buspirone hydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H31 N5 O2 . Cl H
Certifications/Compliance ISO 17034
Synonym Buspirone Hydrochloride
InChI Formula InChI=1S/C21H31N5O2.ClH/c27-18-16-21(6-1-2-7-21)17-19(28)26(18)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20;/h5,8-9H,1-4,6-7,10-17H2;1H
SMILES Cl.O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
Molecular Weight (g/mol) 421.96
Quantity 500 mg
Formula Weight 421.2245 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.