missing translation for 'onlineSavingsMsg'
Learn More

Buspirone hydrochloride, MedChemExpress

Product Code. 16498616
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498616 Supplier Medchem Express Supplier No. HYB1115
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Buspirone hydrochloride is a 5-HT1A receptor agonist. Buspirone hydrochloride can be used for anxiety and depression research.

Buspirone hydrochloride is a 5-HT1A receptor agonist. Buspirone hydrochloride can be used for anxiety and depression research.

Specifications

Chemical Name or Material Buspirone hydrochloride
CAS 33386-08-2
Color White
Purity Grade Notes Research
Molecular Formula C21H32ClN5O2
Quantity 100 mg
Solubility Information H2O : ≥ 100 mg/mL (236.99 mM) ∣DMSO : 50 mg/mL (118.49 mM; Need ultrasonic)
SMILES O=C(N(CCCCN1CCN(C2=NC=CC=N2)CC1)C(C3)=O)CC43CCCC4.[H]Cl
Molecular Weight (g/mol) 421.96
Formula Weight 421.96
Percent Purity 99.56%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.