Learn More
Uvex Busingess Casual 95388 Insole - Width 11
Product Code. 17288171 Shop All Uvex Products
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17288171 39 1 pair
17298171 40 1 pair
17208181 41 1 pair
17218181 42 1 pair
17228181 43 1 pair
17238181 44 1 pair
17248181 45 1 pair
17258181 46 1 pair
17268181 47 1 pair
17278181 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17288171 Supplier Uvex Supplier No. 9538839
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Comfortable climatic insole with antistatic properties for Uvex 1 business Casual
  • Full surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Perforation for improved breathability
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No. 9538.8
  • Size: 38 to 48
TRUSTED_SUSTAINABILITY
Specifikationer
Color Black
For Use With (Equipment) Uvex Business Casual
Material Microvelour
Size 39
Disposable Reusable
Gender Unisex
Fit Standard width
Latex Free Yes
Waterproof No
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.