Learn More
Karl Hecht™ Assistent™ Burette Tips
Product Code. 10217521
Change view
Click to view available options
Type:
Buret Tip
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10217521 Buret Tip Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10217521 Supplier Karl Hecht™ Supplier No. 40780009
  • ONLINE EXCLUSIVE PRICE
    3.35 EUR
    1.83 EUR / Pack of 1
    Save 1.52 EUR
    45% Off
Estimated Shipment: 03-12-2026
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Karl Hecht™ Assistent™ Burette Tips are manufactured from AR-Glas™ soda glass with rubber tubing.
  • Length approx. 60mm
  • For burettes with tubing connection
Specifications
For Use With (Equipment) Burets with tubing connection
Includes Rubber Tubing
Type Buret Tip