Learn More
Fisherbrand™ Bunsen Burner Neoprene Tubing
Product Code. 11855522 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Overall Length (Metric):
450 mm
600 mm
750 mm
900 mm
2 m
10 m
Unit Size:
10m
2m
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) Unit Size
11855522 600 mm Each
12330768 10 m 10m
10492624 2 m 2m
11845522 450 mm Each
11865522 750 mm Each
11875522 900 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11855522
  • ONLINE EXCLUSIVE PRICE
    17.31 EUR
    11.10 EUR / Pack of 1
    Save 6.21 EUR
    35% Off
View all promotions
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
Suitable for use with coal and natural (methane) gas burners.
With reinforced ends for fitting to Bunsen burner and gas tap
Specifications
Material Neoprene
Overall Length (Metric) 600 mm
Thickness (Metric) 1.5 mm
For Use With Bunsen Burner