Learn More
Fisherbrand™ Bulb for Diascopic Light
Product Code. 11532673
Change view
Click to view available options
Voltage:
12 V
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
11532673 12 V Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11532673 Supplier Fisherbrand™ Supplier No. 7260.6441.
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Illuminate Inverso TC100 Microscopes with this light bulb. Fisherbrand™ Bulbs for Diascopic Light are ergonomically designed for everyday use.
  • G4 Halogen Bulb for diascopic light
  • Accessory for microscopes: FB69155 and FB69156
  • Available in packs of ten
Specifications
Voltage 12 V
For Use With (Equipment) diascopic light
Magnification 2.5 to 120x
Shape Diamond
Product Type Bulb