missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution (Boric acid/sodium hydroxide/hydrogen chloride), Supelco™

Product Code. 15815300
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15815300 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815300 Supplier Supelco Supplier No. 1094601000
  • ONLINE EXCLUSIVE PRICE
    78.00 EUR
    56.34 EUR / 1 L
    Save 21.66 EUR
    27% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Traceable to SRM from NIST and PTB pH 8.00 (20°C) Certipur™

The certified buffer solution (boric acid/sodium hydroxide/hydrogen chloride) is gravimetrically prepared using high-quality reagents following DIN EN ISO/IEC 17025. Its pH value of 8.00 at 20°C is measured following a 5-point calibration method, using a combined glass electrode. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. Its pH value is strongly dependent on the temperature and therefore a constant temperature must be maintained during its use. Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.00 g/cm3 at 20°C
For Use With (Application) Calibration and Maintenance
Quantity 1 L
Solution Type Calibration
Grade Research
compliance-icons compliance-icons
Product Identifier
  • Buffer solution pH 8.00 (20 GRAD C) Certipur®
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H290-May be corrosive to metals.
  • H360FD-May damage fertility. May damage the unborn child.
  • EUH208-Contains Mixture of 5-Chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-2H -isothiazol-3-one (3:1). May produce an allergic reaction.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.