Learn More
Buffer concentrate pH 6.00, For 500 ml buffer solution, citric acid / sodium hydroxide solution, Honeywell Fluka™
Product Code. 15635330 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
6 x 1 Ea.
Unit Size:
Pack of 6
Product Code. Quantity unitSize
15635330 6 x 1 Ea. Pack of 6
1 options
Product Code. 15635330
Brand: Honeywell-Fluka 387456X1EA
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Chemical Name or Material Buffer concentrate pH 6.00
Name Note For 500 ml buffer solution, citric acid / sodium hydroxide solution
CAS 100-56-1, 1310-73-2, 7732-18-5, 5949-29-1
CAS Max % ≤0.0010%, ≤9.0000%
CAS Min % ≥7.0000%
Quantity 6 x 1 Ea.
Solution Type Buffer
Packaging Ampoule