Learn More
Buffer, 3.00±0.02, Honeywell Fluka™ Hydrion™
Product Code. 15682400 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15682400 1 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15682400 Supplier Honeywell Fluka Supplier No. 2390111PAK
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
pH 3.00±0.02
  • Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Buffer, Hydrion™
pH 3.00 ±0.02
Quantity 1 Pk.
MDL Number MFCD00132752
Packaging Foil envelope
UN Number NONH for all modes of transport