missing translation for 'onlineSavingsMsg'
Learn More

Buffer, 10.00±0.02, Honeywell Fluka™ Hydrion™

Product Code. 15672410 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15672410 1 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15672410 Supplier Honeywell Fluka Supplier No. 2391271PAK
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

pH 10.00±0.02

  • Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer, Hydrion™
pH 10.00 ±0.02
Quantity 1 Pk.
MDL Number MFCD00132752
Packaging Foil envelope
UN Number NONH for all modes of transport
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.