Learn More
BT-13, MedChemExpress
Product Code. 16495866
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495866 100 mg 100mg
16465168 10 mg 10mg
16407897 5 mg 5mg
16401397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16495866 Supplier Medchem Express Supplier No. HY124401
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress BT-13 is a potent and selective glial cell line-derived neurotrophic factor (GDNF) receptor RET agonist independently of GFLs, promoting neurite growth from sensory neurons in vitro and attenuates experimental neuropathy in the Rat.
BT-13 is a potent and selective glial cell line-derived neurotrophic factor (GDNF) receptor RET agonist independently of GFLs, promoting neurite growth from sensory neurons in vitro and attenuates experimental neuropathy in the Rat.
Specifications
Chemical Name or Material BT-13
CAS 924537-98-4
Color White
Purity Grade Notes Research
Molecular Formula C23H27F4N3O4S
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (48.31 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(OC)C(N2CCN(C(C3=CC=C(F)C=C3C(F)(F)F)=O)CC2)=C1)(N(CC)CC)=O
Molecular Weight (g/mol) 517.54
Formula Weight 517.54
Percent Purity 99.59%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation