Learn More
Thermo Scientific™ Bsu15I (ClaI) (10 U/μL)
Product Code. p-4521566 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1,000 units
3,000 units
600 U
Unit Size:
1000 units
3000 units
600 units
Product Code. 10161140
Brand: Thermo Scientific™ ER0141
Cut at AT^CGAT sites with Bsu15I (ClaI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: BanIII, Bsa29I, BseCI, BshVI, BspDI, BspXI, BsuTUI).
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications
  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10 U/μL
Incubator Temperature 37°C
Quantity 600 U
Product Type Bsu15I (ClaI)