Learn More
Brucine sulfate hydrate, 98% (dry wt.), water <13%
Product Code. 11426390
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11426390 25 g 25g
11436390 100 g 100g
2 options
Product Code. 11426390
Brand: Thermo Scientific Alfa Aesar A19943.14
Request Bulk or Custom Format
CAS: 652154-10-4 | C46H54N4O12S | 887.01 g/mol
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 652154-10-4
Molecular Formula C46H54N4O12S
Molecular Weight (g/mol) 887.01
MDL Number MFCD00013472
InChI Key HCMSIGALSOEZRW-MDQLRNFDSA-N
Synonym brucine sulfate salt
PubChem CID 118797651
IUPAC Name 10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;hydrate
SMILES OS(O)(=O)=O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1
Specifications
Melting Point 191°C to 196°C (decomposition)
Quantity 25 g
UN Number UN2811
Sensitivity Hygroscopic
Merck Index 14,1455
Solubility Information Soluble in water.
Formula Weight 443.52 (Anhydrous)
Percent Purity 98%
Assay (dry wt.), water <13%
Chemical Name or Material Brucine sulfate hydrate