Learn More
Bromuconazole, TRC
Product Code. 30137077
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137077 25 mg 25mg
17943545 10 mg 10mg
17985035 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137077 Supplier Toronto Research Chemicals Supplier No. TRCB69538025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 116255-48-2
Molecular Formula C13 H12 Br Cl2 N3 O
Molecular Weight (g/mol) 377.06
Synonym 2,5-Anhydro-4-bromo-1,3,4-trideoxy-2-C-(2,4-dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)pentitol, 1H-1,2,4-Triazole, 1-[[4-bromo-2-(2,4-dichlorophenyl)tetrahydro-2-furanyl]methyl]-, Bromuconazole, Granit, Granit (pesticide), LS 860263, Vectra
IUPAC Name 1-[[4-bromo-2-(2,4-dichlorophenyl)oxolan-2-yl]methyl]-1,2,4-triazole
SMILES Clc1ccc(c(Cl)c1)C2(Cn3cncn3)CC(Br)CO2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C13H12BrCl2N3O/c14-9-4-13(20-5-9,6-19-8-17-7-18-19)11-2-1-10(15)3-12(11)16/h1-3,7-9H,4-6H2
Formula Weight 374.9541
Chemical Name or Material Bromuconazole