missing translation for 'onlineSavingsMsg'
Learn More

Bromotrichloromethane, Sigma-Aldrich™

Product Code. 15828700
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15828700 250 mL 250mL
15818700 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15828700 Supplier Sigma Aldrich Supplier No. 8019860250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-62-7
Grade Research
Molecular Weight (g/mol) 198.27

Specifications

Melting Point -6°C
Density 2.01 g/cm3 at 20°C
Boiling Point 105°C/1013 hPa
MDL Number MFCD00000783
Quantity 250 mL
Synonym Bromotrichloromethane, Trichlorobromomethane, Bromotrichloromethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.