Learn More
Bromosuccinic acid, 96%
Product Code. 11469857
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11469857 5 g 5g
11479857 25 g 25g
11489857 100 g 100g
3 options
Product Code. 11469857
Brand: Thermo Scientific Alfa Aesar B22829.06
Request Bulk or Custom Format
CAS: 923-06-8 | C4H5BrO4 | 196.98 g/mol
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.
Solubility
Insoluble in ether.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 923-06-8
Molecular Formula C4H5BrO4
Molecular Weight (g/mol) 196.98
MDL Number MFCD00004213
InChI Key QQWGVQWAEANRTK-UHFFFAOYNA-N
Synonym bromosuccinic acid, 2-bromosuccinic acid, monobromosuccinic acid, butanedioic acid, bromo, +--bromosuccinic acid, dl-bromosuccinic acid, rs-bromosuccinic acid, succinic acid, bromo, bromobutanedioic acid
PubChem CID 73557
ChEBI CHEBI:73712
IUPAC Name 2-bromobutanedioic acid
SMILES OC(=O)CC(Br)C(O)=O
Specifications
Melting Point 164°C to 166°C (decomposition)
Quantity 5 g
Beilstein 1723556
Merck Index 14,1437
Solubility Information Insoluble in ether.
Formula Weight 196.99
Percent Purity 96%
Chemical Name or Material Bromosuccinic acid