accessibility menu, dialog, popup

UserName

CAS RN 923-06-8

HOOBrOHO

CAS RN 923-06-8

IUPAC Name: 2-bromobutanedioic acid
Synonyms: bromosuccinic acid
Molecular Weight (g/mol): 196.98
Molecular Formula: C4H5BrO4
InChi Key: QQWGVQWAEANRTK-UHFFFAOYNA-N
SMILES: OC(=O)CC(Br)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results