Learn More
Bromonitromethane, TRC
Specifications
Specifications
| CAS | 563-70-2 |
| Molecular Formula | C H2 Br N O2 |
| Quantity | 25 g |
| Synonym | Bromonitromethane, Nitrobromomethane, α-Bromonitromethane; |
| InChI Formula | InChI=1S/CH2BrNO2/c2-1-3(4)5/h1H2 |
| SMILES | [O-][N+](=O)CBr |
| IUPAC Name | bromo(nitro)methane |
| Molecular Weight (g/mol) | 139.94 |
| Formula Weight | 138.9269 |
| Chemical Name or Material | Bromonitromethane |