Learn More
Bromoform, Sigma-Aldrich™
Product Code. 15870561
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870561 1 L 1 L
15860561 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15870561 Supplier Sigma Aldrich Supplier No. 8187021000
 Restricted Item
Estimated Shipment: 16-11-2026
Stabilized with ethanol for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-25-2
Grade Research
Molecular Weight (g/mol) 252.73
Specifications
Melting Point 9.2°C
Density 2.89 g/cm3 at 20°C
Boiling Point 149.5°C/1013 mbar
MDL Number MFCD00000128
Quantity 1 L
Synonym Bromoform, Tribromomethane, Methane tribromide, Methyl tribromide, Bromoform