missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, Supelco™

Product Code. 15895090
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15895090 1 g 1g
15805100 5 g 5g
15815100 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15895090 Supplier Supelco Supplier No. 1081210001
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-60-8
Grade Research
Molecular Weight (g/mol) 698.01

Specifications

Melting Point 217°C to 218°C
MDL Number MFCD00005874
Synonym Bromocresol green, 3,3′,5,5′-Tetrabromo-m-cresolsulfonphthalein, BCG, Bromocresol green
Quantity 1 g
Chemical Name or Material Bromocresol Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.