Learn More
Bromochloroacetamide, TRC
Product Code. 30134257
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134257 1 g 1g
30137902 250 mg 250mg
17994675 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134257 Supplier Toronto Research Chemicals Supplier No. TRCB7400481G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62872-34-8
Molecular Formula C2H3BrClNO
Molecular Weight (g/mol) 172.41
Synonym 2-Bromo-2-chloroacetamide;
IUPAC Name 2-bromo-2-chloroacetamide
SMILES NC(=O)C(Cl)Br
Specifications
Quantity 1 g
InChI Formula InChI=1S/C2H3BrClNO/c3-1(4)2(5)6/h1H,(H2,5,6)
Formula Weight 170.9087
Percent Purity >95%
Chemical Name or Material Bromochloroacetamide
Grade HPLC