Learn More
R&D Systems™ BromoCatch™ GFP Expression Plasmid (C-term, pCMV, Puro)
Product Code. 31900335
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31900335 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31900335 Supplier R&D Systems™ Supplier No. RDEHBC02GFP050
 Restricted Item
Estimated Shipment: 30-11-2026
This plasmid enables expression of proteins fused to the BromoCatch™ tag, a covalent self-labeling tag derived from a BET bromodomain.
TRUSTED_SUSTAINABILITY
Specifications
Product Line R&D Systems
Quantity 50 μg