missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic-1,2-13C2 Acid, TRC

Product Code. 30012603
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012603 250 mg 250mg
30012192 1 g 1g
30006314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012603 Supplier Toronto Research Chemicals Supplier No. TRCB679078250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52947-00-9
Molecular Formula 13C2 H3 Br O2
Molecular Weight (g/mol) 140.93
Synonym Acetic-13C2 acid, bromo- (9CI), [1,2-13C2]Bromoacetic acid, Bromoacetic-1,2-13C acid, 2-Bromoacetic-1,2-13C2 acid, Bromoacetic Acid-13C2
IUPAC Name 2-bromoacetic acid
SMILES O[13C](=O)[13CH2]Br

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)/i1+1,2+1
Formula Weight 139.9384
Chemical Name or Material Bromoacetic acid-13C2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.