missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic-1-13C Acid, TRC

Product Code. 30008561
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008561 500 mg 500mg
30007931 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008561 Supplier Toronto Research Chemicals Supplier No. TRCB679077500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57858-24-9
Molecular Formula 13C C H3 Br O2
Molecular Weight (g/mol) 139.94
Synonym Acetic-1-13C acid, 2-bromo- (9CI), Bromoacetic acid-1-13C, 2-Bromoacetic-1-13C acid, Bromoacetic-1-13C Acid
IUPAC Name 2-bromoacetic acid
SMILES O[13C](=O)CBr

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)/i2+1
Formula Weight 138.935
Percent Purity >95%
Chemical Name or Material Bromoacetic-1-13C Acid
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.