Learn More
Bromine, Supelco™
Product Code. 15887040
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15887040 1 L 1 L
15877040 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15887040 Supplier Supelco Supplier No. 1019451000
 Restricted Item
Estimated Shipment: 19-11-2026
Extra pure
extra pure
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7726-95-6
Grade Research
Molecular Weight (g/mol) 159.81
Specifications
Boiling Point 58.8°C/1013 hPa
Melting Point -7.3°C
Quantity 1 L
MDL Number MFCD00010896
Synonym Bromine, Bromine
Density 3.12 g/cm3 at 20°C (Liquid)