missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Ponceau G, MP Biomedicals

Product Code. 11467735 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467735 10 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11467735 Supplier MP Biomedicals Supplier No. 0521433901

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3761-53-3
Molecular Formula C18H14N2Na2O7S2
InChI Key YJVBLROMQZEFPA-UHFFFAOYSA-L
SMILES CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
IUPAC Name disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
Molecular Weight (g/mol) 480.417
PubChem CID 102092631
ChEBI CHEBI:82369
Formula Weight 480.4
Quantity 10 g
Chemical Name or Material Brilliant Ponceau G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.