Learn More
Brij™ 98
Product Code. 10641832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10641832 500 g Glass bottle 500g
10117553 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10641832 Supplier Thermo Scientific Acros Supplier No. 347185000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
CAS: 9004-98-2 | C58H116O21 | 1149.55 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9004-98-2
Molecular Formula C58H116O21
Molecular Weight (g/mol) 1149.55
MDL Number MFCD00074369
InChI Key TVSFHLHLHUYZNN-KTKRTIGZSA-N
Synonym Polyoxyethylene(20) oleyl ether
PubChem CID 5364713
IUPAC Name (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
Specifications
Color White to Cream
Acid Value 1mg KOH/g max.
Physical Form Soft Waxy Solid
Boiling Point >100.0°C
Melting Point 30.0°C
Density 1.0700g/mL
Quantity 500 g
Flash Point >150°C
Hydroxyl Value 50 to 65mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.07
Linear Formula C18H35(OCH2CH2)nOH
Water 3% max.
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, toluene, propylene glycol, insoluble in cottonseed oil, mineral oil
Formula Weight 1149.56
Chemical Name or Material Brij™ 98
Show More Show Less
compliance-icons
Product Identifier
  • Brij|TS 98
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.