Learn More
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits
Product Code. 17827071
Change view
Click to view available options
Description:
CFP BrightComp eBeads Compensation Bead Kit
GFP BrightComp eBeads Compensation Bead Kit
RFP BrightComp eBeads Compensation Bead Kit
YFP BrightComp eBeads Compensation Bead Kit
mCherry BrightComp eBeads Compensation Bead Kit
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
17827071 mCherry BrightComp eBeads Compensation Bead Kit Pack of 25
17897061 RFP BrightComp eBeads Compensation Bead Kit Pack of 25
17807071 YFP BrightComp eBeads Compensation Bead Kit Pack of 25
17817071 CFP BrightComp eBeads Compensation Bead Kit Pack of 25
15980213 GFP BrightComp eBeads Compensation Bead Kit Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17827071 Supplier Invitrogen™ Supplier No. A54743
  • ONLINE EXCLUSIVE PRICE
    285.00 EUR
    213.65 EUR / Pack of 25
    Save 71.35 EUR
    25% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
• Three levels of intensity, perfect for any samples no matter the level of expression
• Near-identical, spectrally-matched dye to fluorescent protein
• Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
TRUSTED_SUSTAINABILITY
Specifications
Description mCherry BrightComp eBeads Compensation Bead Kit
Quantity 25 tests
Excitation Wavelength Range 561 nm
For Use With (Application) Flow Cytometry
Product Type Compensation Beads