Learn More
Hirschmann™ Bright Lined Counting Chambers
Product Code. 10283861
Change view
Click to view available options
Product Type:
Counting Chamber (Burker)
Counting Chamber (Fuch-Rosenthal)
Counting Chamber (Neubauer)
Counting Chamber (Thoma)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10283861 Counting Chamber (Burker) Each
10469132 Counting Chamber (Neubauer) Each
10517831 Counting Chamber (Thoma) Each
10528961 Counting Chamber (Fuch-Rosenthal) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10283861 Supplier Hirschmann™ Supplier No. 8100201
  • ONLINE EXCLUSIVE PRICE
    169.00 EUR
    149.00 EUR / Pack of 1
    Save 20.00 EUR
    11% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Includes 2 haemocytometer glass covers.
  • Double net ruling
  • Bright lined
  • With LOT number on packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
Product Type Counting Chamber (Burker)
Depth (Metric) 0.1 mm
Certifications/Compliance CE pursuant to IVD 98/79 EC
Includes 2 haemocytometer glass covers