Learn More
MP Biomedicals™ Briggs N Salt Mixture, Powder
Product Code. 11425632 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425632 50 kg 50kg
11405632 2 kg 2kg
11415632 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11425632 Supplier MP Biomedicals™ Supplier No. 0290283450
  • ONLINE EXCLUSIVE PRICE
    3490.00 EUR
    3320.10 EUR / 50kg
    Save 169.90 EUR
    4% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Salt mixture for research animal diet.
  • Salt Mixture
  • For research animal diet
  • Powder
Specifications
Product Type Animal Diet
For Use With (Application) Reseach animal diet
Quantity 50 kg