missing translation for 'onlineSavingsMsg'
Learn More

Brexpiprazole, TRC

Product Code. 30008061
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008061 100 mg 100mg
30003533 50 mg 50mg
30007834 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008061 Supplier Toronto Research Chemicals Supplier No. TRCB677385100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 913611-97-9
Molecular Formula C25 H27 N3 O2 S
Molecular Weight (g/mol) 433.57
Synonym 2(1H)-Quinolinone, 7-[4-(4-benzo[b]thien-4-yl-1-piperazinyl)butoxy]-, 7-[4-(4-Benzo[b]thien-4-yl-1-piperazinyl)butoxy]-2(1H)-quinolinone, 7-[4-(4-(Benzo[b]thien-4-yl)-piperazin-1-yl)butoxy]-1H-quinolin-2-one, Brexpiprazole, OPC 34712, Rexulti, Rxulti
IUPAC Name 7-[4-[4-(1-benzothiophen-4-yl)piperazin-1-yl]butoxy]-1H-quinolin-2-one
SMILES O=C1Nc2cc(OCCCCN3CCN(CC3)c4cccc5sccc45)ccc2C=C1

Specifica

Quantity 100 mg
InChI Formula InChI=1S/C25H27N3O2S/c29-25-9-7-19-6-8-20(18-22(19)26-25)30-16-2-1-11-27-12-14-28(15-13-27)23-4-3-5-24-21(23)10-17-31-24/h3-10,17-18H,1-2,11-16H2,(H,26,29)
Formula Weight 433.1824
Percent Purity >95%
Chemical Name or Material Brexpiprazole
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.